Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butynyl p-toluenesulfonate, 98%

Chemical Identifiers

Chemical Name or Material 2-Butynyl p-toluenesulfonate
CAS 56563-37-2
Molecular Formula C11H12O3S
Formula Weight 224.28g/mol
MDL Number MFCD00078395
Beilstein 3300477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1249903

 
alfa aesar™
L12499-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 47° to 48°C
Quantity 1g
Chemical Name or Material 2-Butynyl p-toluenesulfonate
CAS 56563-37-2
Assay 98%
Molecular Formula C11H12O3S
Formula Weight 224.28g/mol
MDL Number MFCD00078395
Beilstein 3300477