Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butynyl acetate, 98%

Chemical Identifiers

Chemical Name or Material 2-Butynyl acetate
CAS 34485-37-5
Molecular Formula CH3CO2CH2C≡CCH3
Formula Weight 112.13
MDL Number MFCD00078282
UN Number UN3272
Beilstein 1745269
Synonym Acetic acid 2-butynyl ester; 1-Acetoxy-2-butyne
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2458406

 
alfa aesar™
B2458406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.13
Chemical Name or Material 2-Butynyl acetate
Assay 98%
Molecular Formula CH3CO2CH2C≡CCH3
MDL Number MFCD00078282
CAS 34485-37-5
UN Number UN3272
Beilstein 1745269
Synonym Acetic acid 2-butynyl ester; 1-Acetoxy-2-butyne
Health Hazard 1 Warning
Boiling Point 150° to 152°C (680mmHg)
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Quantity 5g
Flash Point 56°C (132°F)
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.