Learn More
2-Butyn-1-ol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 223400050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Butyn-1-ol | |
764-01-2 | |
100.0 | |
142.0°C to 143.0°C | |
Authentic | |
Glass bottle | |
1.4520 to 1.4540 | |
MFCD00002914 | |
01, II, 505 | |
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
CC#CCO | |
70.09 | |
70.09 |
95% | |
97.5 | |
0.9300g/mL | |
51°C | |
97.5% min. (GC) | |
C4H6O | |
CH3C≡CCH2OH | |
5mL | |
0.93 | |
NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
but-2-yn-1-ol | |
12991 | |
98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: R
GHS Signal Word: Warning
EINECSNumber : 212-113-4
TSCA : TSCA
RUO â Research Use Only