missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butyn-1-ol, 98%

Manufacturer:  Alfa Aesar™ A1570906

 View more versions of this product

Catalog No. AAA1570906



Description

Description

Applications
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

Solubility
Miscible with chloroform and methanol.

Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
Specifications

Specifications

2-Butyn-1-ol
Characteristic
-2°C
764-01-2
MFCD00002914
1733676
NEEDEQSZOUAJMU-UHFFFAOYSA-N
but-2-yn-1-ol
12991
98%
51°C (123°F)
142°C to 143°C
5g
C4H6O
UN1987
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
CC#CCO
70.091
70.09

Chemical Identifiers

764-01-2
70.091
NEEDEQSZOUAJMU-UHFFFAOYSA-N
12991
CC#CCO
C4H6O
MFCD00002914
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
but-2-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

Documents