Learn More
2-Butyn-1-ol, 98%, Thermo Scientific Chemicals
$90.75 - $995.48
Chemical Identifiers
CAS | 764-01-2 |
---|---|
Molecular Formula | C4H6O |
Molecular Weight (g/mol) | 70.091 |
MDL Number | MFCD00002914 |
InChI Key | NEEDEQSZOUAJMU-UHFFFAOYSA-N |
Synonym | 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol |
PubChem CID | 12991 |
IUPAC Name | but-2-yn-1-ol |
SMILES | CC#CCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1570906
|
Thermo Scientific Chemicals
A1570906 |
5 g |
Each for $90.75
|
|
|||||
AAA1570914
|
Thermo Scientific Chemicals
A1570914 |
25 g |
Each for $301.73
|
|
|||||
AAA1570922
|
Thermo Scientific Chemicals
A1570922 |
100 g |
Each for $995.48
|
|
|||||
Description
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.
Solubility
Miscible with chloroform and methanol.
Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
Chemical Identifiers
764-01-2 | |
70.091 | |
NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
12991 | |
CC#CCO |
C4H6O | |
MFCD00002914 | |
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
but-2-yn-1-ol |
Specifications
764-01-2 | |
0.938 | |
51°C (123°F) | |
C4H6O | |
MFCD00002914 | |
UN1987 | |
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
but-2-yn-1-ol | |
12991 | |
98% |
-2°C | |
142°C to 143°C | |
Characteristic | |
1.453 | |
5 g | |
1733676 | |
Miscible with chloroform and methanol. | |
CC#CCO | |
70.091 | |
70.09 | |
2-Butyn-1-ol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 212-113-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only