Learn More
2-Butyn-1-ol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1570922
Description
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.
Solubility
Miscible with chloroform and methanol.
Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
Specifications
2-Butyn-1-ol | |
-2°C | |
142°C to 143°C | |
Characteristic | |
1.453 | |
100 g | |
1733676 | |
Miscible with chloroform and methanol. | |
CC#CCO | |
70.091 | |
70.09 |
764-01-2 | |
0.938 | |
51°C (123°F) | |
C4H6O | |
MFCD00002914 | |
UN1987 | |
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
but-2-yn-1-ol | |
12991 | |
98% |
Chemical Identifiers
764-01-2 | |
70.091 | |
NEEDEQSZOUAJMU-UHFFFAOYSA-N | |
12991 | |
CC#CCO |
C4H6O | |
MFCD00002914 | |
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol | |
but-2-yn-1-ol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 212-113-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only