missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butyn-1-al diethyl acetal, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 223410010

 View more versions of this product

Catalog No. AC223410010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Butyn-1-al diethyl acetal
2806-97-5
100.0
Liquid
98%
CH3C2CCH(OC2H5)2
142.2
01, 750
1.4250 to 1.4270 (20°C, 589nm)
Clear light yellow to yellow
98%
97.5
Authentic
≥97.5%
C8H14O2
Glass bottle
MFCD00009274
15,66
1, 1-Diethoxy-2-butyne
1g

Chemical Identifiers

2806-97-5
100.0
C8H14O2
142.2
1, 1-Diethoxy-2-butyne
≥97.5%
97.5
98%
CH3C2CCH(OC2H5)2
MFCD00009274
2-Butyn-1-al diethyl acetal, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA