missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butyn-1-al diethyl acetal, 98%, ACROS Organics™

Chemical Identifiers

CAS 2806-97-5
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.2
MDL Number MFCD00009274
InChI Key FKKVKKSEVMQYER-UHFFFAOYSA-N
Synonym 1,1-diethoxy-2-butyne #, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, 2-butyn-1-al diethyl acetal, 2-butynal diethyl acetal, 2-butyne, 1,1-diethoxy, 2-butynyl aldehyde diethyl acetal, acmc-20ao12, tetrolaldehyde diethyl acetal
PubChem CID 137721
IUPAC Name 1,1-diethoxybut-2-yne
SMILES CCOC(C#CC)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC223410010
SDS View Documents
Thermo Fisher Scientific
223410010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2806-97-5
142.2
FKKVKKSEVMQYER-UHFFFAOYSA-N
137721
CCOC(C#CC)OCC
C8H14O2
MFCD00009274
1,1-diethoxy-2-butyne #, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, 2-butyn-1-al diethyl acetal, 2-butynal diethyl acetal, 2-butyne, 1,1-diethoxy, 2-butynyl aldehyde diethyl acetal, acmc-20ao12, tetrolaldehyde diethyl acetal
1,1-diethoxybut-2-yne

Specifications

0.9000g/mL
Glass bottle
0.9
Yellow
2806-97-5
100.0
C8H14O2
MFCD00009274
15,66
FKKVKKSEVMQYER-UHFFFAOYSA-N
1,1-diethoxybut-2-yne
137721
Liquid
2-Butyn-1-al diethyl acetal, 98%
Authentic
1.4250 to 1.4270
65.0°C to 67.0°C (12.0mmHg)
1g
97.5
98%
CH3C≡CCH(OC2H5)2
01, 750
1,1-diethoxy-2-butyne #, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, 2-butyn-1-al diethyl acetal, 2-butynal diethyl acetal, 2-butyne, 1,1-diethoxy, 2-butynyl aldehyde diethyl acetal, acmc-20ao12, tetrolaldehyde diethyl acetal
CCOC(C#CC)OCC
142.2
142.2
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS