missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butylbenzofuran, TCI America™

$117.64 - $947.23

Chemical Identifiers

CAS 4265-27-4
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00005851
InChI Key OVJKFJDEVKABNF-UHFFFAOYSA-N
Synonym 2-butylbenzofuran, benzofuran, 2-butyl, 2-n-butylbenzo b furan, 2-butyl-benzofuran, 2-n-butylbenzofuran, 2-butylbenzo b furan, butylbenzofuran, 2-n-butylbezofuran, butyl-2-benzofuran, 2-n-butylbenzofurane
PubChem CID 77951
IUPAC Name 2-butyl-1-benzofuran
SMILES CCCCC1=CC2=CC=CC=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B182425G View Documents TCI America
B182425G
25g Each for $117.64
 
B1824500G View Documents TCI America
B1824500G
500g Each for $947.23
 
Specifications

Chemical Identifiers

4265-27-4
174.243
OVJKFJDEVKABNF-UHFFFAOYSA-N
77951
CCCCC1=CC2=CC=CC=C2O1
C12H14O
MFCD00005851
2-butylbenzofuran, benzofuran, 2-butyl, 2-n-butylbenzo b furan, 2-butyl-benzofuran, 2-n-butylbenzofuran, 2-butylbenzo b furan, butylbenzofuran, 2-n-butylbezofuran, butyl-2-benzofuran, 2-n-butylbenzofurane
2-butyl-1-benzofuran

Specifications

129°C
4265-27-4
MFCD00005851
OVJKFJDEVKABNF-UHFFFAOYSA-N
2-butyl-1-benzofuran
77951
≥99.0% (GC)
25g
C12H14O
2-butylbenzofuran, benzofuran, 2-butyl, 2-n-butylbenzo b furan, 2-butyl-benzofuran, 2-n-butylbenzofuran, 2-butylbenzo b furan, butylbenzofuran, 2-n-butylbezofuran, butyl-2-benzofuran, 2-n-butylbenzofurane
CCCCC1=CC2=CC=CC=C2O1
174.243
174.24
2-Butylbenzofuran
Safety and Handling

Safety and Handling

TSCA : No