Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Butyl)triphenylphosphonium bromide, 96%

$157.00 - $157.00

Chemical Identifiers

Chemical Name or Material (2-Butyl)triphenylphosphonium bromide
CAS 3968-92-1
Molecular Formula C22H24BrP
Formula Weight 399.32
MDL Number MFCD00050236
Beilstein 3580097
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0153206

 
alfa aesar™
L0153206
5g Each for $157.00
Description & Specifications

Specifications

Formula Weight 399.32
Chemical Name or Material (2-Butyl)triphenylphosphonium bromide
Molecular Formula C22H24BrP
CAS 3968-92-1
Beilstein 3580097
Assay 96%
MDL Number MFCD00050236
Health Hazard 1 Warning
Melting Point 227° to 237°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.