missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butyl-2-ethyl-1,3-propanediol, TCI America™

$39.72 - $189.45

Chemical Identifiers

CAS 115-84-4
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.257
MDL Number MFCD00004697
InChI Key DSKYSDCYIODJPC-UHFFFAOYSA-N
Synonym 2-butyl-2-ethyl-1,3-propanediol, 2-ethyl-2-butyl-1,3-propanediol, 3,3-bis hydroxymethyl heptane, 2-butyl-2-ethylpropanediol, caswell no. 129, 1,3-propanediol, 2-butyl-2-ethyl, 2-ethyl-2-butyl-1,3-propandeiol, bepd, unii-61p3kj8k4y, bep
PubChem CID 61038
IUPAC Name 2-butyl-2-ethylpropane-1,3-diol
SMILES CCCCC(CC)(CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B0922500G View Documents TCI America
B0922500G
500g Each for $189.45
 
B092225G View Documents TCI America
B092225G
25g Each for $39.72
 
Specifications

Chemical Identifiers

115-84-4
160.257
DSKYSDCYIODJPC-UHFFFAOYSA-N
61038
CCCCC(CC)(CO)CO
C9H20O2
MFCD00004697
2-butyl-2-ethyl-1,3-propanediol, 2-ethyl-2-butyl-1,3-propanediol, 3,3-bis hydroxymethyl heptane, 2-butyl-2-ethylpropanediol, caswell no. 129, 1,3-propanediol, 2-butyl-2-ethyl, 2-ethyl-2-butyl-1,3-propandeiol, bepd, unii-61p3kj8k4y, bep
2-butyl-2-ethylpropane-1,3-diol

Specifications

178°C
43°C
115-84-4
MFCD00004697
DSKYSDCYIODJPC-UHFFFAOYSA-N
2-butyl-2-ethylpropane-1,3-diol
61038
Crystal-Powder at 20°C
2-Butyl-2-ethyl-1,3-propanediol
White
500g
C9H20O2
2-butyl-2-ethyl-1,3-propanediol, 2-ethyl-2-butyl-1,3-propanediol, 3,3-bis hydroxymethyl heptane, 2-butyl-2-ethylpropanediol, caswell no. 129, 1,3-propanediol, 2-butyl-2-ethyl, 2-ethyl-2-butyl-1,3-propandeiol, bepd, unii-61p3kj8k4y, bep
CCCCC(CC)(CO)CO
160.257
160.26
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0240&(2)-2391

RTECSNumber : TY3605000

TSCA : Yes