missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butyl-2-adamantanol, TCI America™

$209.31 - $640.16

Chemical Identifiers

CAS 14451-86-6
Molecular Formula C14H24O
Molecular Weight (g/mol) 208.345
InChI Key PXRAOOXXPGRCJZ-UHFFFAOYSA-N
Synonym 2-Butyl-2-hydroxyadamantane
PubChem CID 12627253
IUPAC Name 2-butyladamantan-2-ol
SMILES CCCCC1(C2CC3CC(C2)CC1C3)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28395G View Documents TCI America
B28395G
5g Each for $209.31
 
B283925G View Documents TCI America
B283925G
25g Each for $640.16
 
Specifications

Chemical Identifiers

14451-86-6
208.345
2-Butyl-2-hydroxyadamantane
2-butyladamantan-2-ol
C14H24O
PXRAOOXXPGRCJZ-UHFFFAOYSA-N
12627253
CCCCC1(C2CC3CC(C2)CC1C3)O

Specifications

White-Yellow
5g
C14H24O
PXRAOOXXPGRCJZ-UHFFFAOYSA-N
2-butyladamantan-2-ol
12627253
Crystal-Powder at 20°C
2-Butyl-2-adamantanol
79°C
14451-86-6
2-Butyl-2-hydroxyadamantane
CCCCC1(C2CC3CC(C2)CC1C3)O
208.345
208.35
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No