Please login to your online account to display your discounted pricing

2-Butoxyethanol, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$62.99 - $206.74

Chemical Identifiers

Chemical Name or Material 2-Butoxyethanol
CAS 111-76-2
Molecular Formula CH3(CH2)3OCH2CH2OH
Formula Weight 118.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002076

 
avantor performance
D64809
Narrow Mouth Amber Glass Bottle 4L Each for $206.74

02002077

 
avantor performance
D64807
Narrow Mouth Amber Glass Bottle 500mL Each for $62.99
Description & Specifications

Specifications

Formula Weight 118.18
CAS 111-76-2
Chemical Name or Material 2-Butoxyethanol
Molecular Formula CH3(CH2)3OCH2CH2OH
Flash Point 62.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.90kg
Quantity 4L