Please login to your online account to display your discounted pricing

2-Butoxyethanol, 99%, ACROS Organics™


Quantity 1L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  154330010

 View more versions of this product

Catalog No. AC154330010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Purity Grade Extra Pure
Assay ≥98.5%
Formula Weight 118.18
Quantity 1L
Color Clear
Beilstein 01, II, 519
Merck Index 15, 1560
Packaging Glass Bottle
CAS Max % 100.0
Refractive Index 1.4180 to 1.4220 (20°C, 589nm)
Linear Formula CH3(CH2)3OCH2CH2OH
Residue after Evaporation 0.02% max.
Infrared Spectrum Authentic
MDL Number MFCD00002884
CAS 111-76-2
Chemical Name or Material 2-Butoxyethanol
CAS Min % 98.5
Health Hazard 1 Warning
Density (20°C) 0.899 to 0.903kg/L
Water 0.1% max. (Coulometric)
Synonym Butyl Cellosolve™
Acetic Acid 0.01% max.
Molecular Formula C6H14O2
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Butoxyethanol
CAS 111-76-2
CAS Min % 98.5
CAS Max % 100.0
Purity Grade Extra Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14O2
Linear Formula CH3(CH2)3OCH2CH2OH
Formula Weight 118.18
MDL Number MFCD00002884
Synonym Butyl Cellosolve™
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents