missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butene-1,4-bis(triphenylphosphonium chloride), 98+%

$72.16 - $272.96

Chemical Identifiers

CAS 106423-29-4
Molecular Formula C40H36Cl2P2
Molecular Weight (g/mol) 649.576
MDL Number MFCD00031713
InChI Key OOACLPDZHDFCQP-UHFFFAOYSA-L
Synonym 2-butene-1,4-bis triphenylphosphonium chloride, trans-2-butene-1,4-bis triphenylphosphoniumchloride, but-2-ene-1,4-diyl bis triphenylphosphanium dichloride
PubChem CID 70700154
IUPAC Name triphenyl(4-triphenylphosphaniumylbut-2-enyl)phosphanium;dichloride
SMILES C1=CC=C(C=C1)[P+](CC=CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0053409 View Documents Alfa Aesar™
L0053409
10g Each for $72.16
 
AAL0053418 View Documents Alfa Aesar™
L0053418
50g Each for $272.96
 
Specifications

Chemical Identifiers

106423-29-4
649.576
OOACLPDZHDFCQP-UHFFFAOYSA-L
70700154
C1=CC=C(C=C1)[P+](CC=CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-].[Cl-]
C40H36Cl2P2
MFCD00031713
2-butene-1,4-bis triphenylphosphonium chloride, trans-2-butene-1,4-bis triphenylphosphoniumchloride, but-2-ene-1,4-diyl bis triphenylphosphanium dichloride
triphenyl(4-triphenylphosphaniumylbut-2-enyl)phosphanium;dichloride

Specifications

264°C to 267°C
106423-29-4
MFCD00031713
2-butene-1,4-bis triphenylphosphonium chloride, trans-2-butene-1,4-bis triphenylphosphoniumchloride, but-2-ene-1,4-diyl bis triphenylphosphanium dichloride
C1=CC=C(C=C1)[P+](CC=CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-].[Cl-]
649.576
649.6
2-Butene-1,4-bis(triphenylphosphonium chloride)
10g
C40H36Cl2P2
3866753
OOACLPDZHDFCQP-UHFFFAOYSA-L
triphenyl(4-triphenylphosphaniumylbut-2-enyl)phosphanium;dichloride
70700154
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning