Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Buten-1-ol, cis + trans (ca 1:19), 96%

$59.72 - $59.72

Chemical Identifiers

Chemical Name or Material 2-Buten-1-ol
CAS 6117-91-5
Name Note cis + trans (≈ 1:19)
Molecular Formula CH3CH=CHCH2OH
Formula Weight 72.11
MDL Number MFCD00002923
UN Number UN1987
Beilstein 1719374
Merck Index 14,2601
Synonym Crotyl alcohol; 1-Hydroxybut-2-ene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1068114

 
alfa aesar™
A1068114
25g Each for $59.72

AAA1068122

 
alfa aesar™
A1068122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1068136

 
alfa aesar™
A1068136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 72.11
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Buten-1-ol
TSCA TSCA
Name Note cis + trans (≈ 1:19)
Assay 96%
Molecular Formula CH3CH=CHCH2OH
MDL Number MFCD00002923
CAS 6117-91-5
UN Number UN1987
Beilstein 1719374
Merck Index 14,2601
Synonym Crotyl alcohol; 1-Hydroxybut-2-ene
Boiling Point 121° to 122°C
Quantity 25g
Flash Point 37°C (98°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302-H312
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number EM9275000

Applications
2-Buten-1-ol, cis + trans (ca 1:19) is used as a building block for the synthesis of various pharmaceutical compounds such as crotyl mercaptan.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides, acid anhydrides and phosphorus halides.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302-H312
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.