Learn More
2-Butanone, for HPLC, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389570025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
2-Butanone | |
78-93-3 | |
0.0003 meq/g max. | |
80.0°C | |
−7°C | |
Liquid | |
0.001% max. | |
C4H8O | |
MFCD00011648 | |
0.1% max. | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
butan-2-one | |
6569 | |
72.11 | |
HPLC |
-87.0°C | |
0.009 max. (1 cm cell vs water) at 350nm, 0.009 max. (1 cm cell vs water) at 450nm, 0.097 max. (1 cm cell vs water) at 340nm, 0.699 max. (1 cm cell vs water) at 330nm, 1 max. (1 cm cell vs water) at 329nm | |
0.8060g/mL | |
2.5 L | |
Glass Bottle | |
1.3780 to 1.3800 | |
0.806 | |
C2H5COCH3 | |
0.42 mPa.s (15°C) | |
15, 6143 | |
Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils | |
CCC(C)=O | |
72.11 | |
CHEBI:28398 | |
99.7% |
Chemical Identifiers
78-93-3 | |
72.11 | |
ZWEHNKRNPOVVGH-UHFFFAOYSA-N | |
6569 | |
butan-2-one |
C4H8O | |
MFCD00011648 | |
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon | |
CHEBI:28398 | |
CCC(C)=O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only