missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butanone, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC396951000

 View more versions of this product

Catalog No. AC396951000


 

Specifications Description & Specifications

Specifications

2-Butanone
78-93-3
100.0
Extra Dry
≥99.5%
C4H8O
72.11
15, 6143
0.8060g/mL
0.001% max.
Clear colorless
Extra Dry
99.4
Filtered over 0.2 micrometer
Liquid
99.5%
C2H5COCH3
MFCD00011648
MEK
AcroSeal™ Glass Bottle
0.005% Max. (Coulometric)
100mL

Chemical Identifiers

78-93-3
100.0
C4H8O
72.11
MEK
≥99.5%
99.4
99.5%
C2H5COCH3
MFCD00011648
2-Butanone, Extra Dry
Extra Dry

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger