missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Butanone, 97%

Manufacturer:  Alfa Aesar™ 31069M1

 View more versions of this product

Catalog No. AA31069M1



Specifications

Specifications

2-Butanone
C4H8O
MFCD00011648
741880
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CCC(=O)C
72.107
CHEBI:28398
Liquid
0.805g/mL
1.3790
80°C
-87°C
78-93-3
CH3COCH2CH3
UN1193
14,6072
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
6569
72.11
97%
Characteristic
−3° (26°F)
Colorless
4 x 1L

Chemical Identifiers

78-93-3
72.107
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
6569
butan-2-one
C4H8O
MFCD00011648
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CHEBI:28398
CCC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL METHYL KETONE

RTECSNumber : EL6475000

TSCA : TSCA

SDS Documents