Learn More
2-Butanol, anhydrous, 99%, Thermo Scientific Chemicals
Hygroscopic; H2O <0.01%
Supplier: Thermo Scientific Chemicals 043315AK
Description
2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Butanol | |
H2O <0.01% | |
Colorless | |
0.808 g/cm3 at 20°C | |
250 mL | |
390°C (734°F) | |
Liquid | |
99% | |
MFCD00004569 | |
1718765 | |
Miscible with methanol and diethyl ether. Slightly miscible with water. | |
CCC(C)O | |
74.12 | |
CHEBI:35687 |
−115°C | |
78-92-2 | |
Colorless liquid | |
98°C to 101°C | |
26°C (79°F) | |
Alcohol-like | |
1.397 | |
C4H10O | |
17.3 hPa (13mm Hg) at 20°C | |
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
butan-2-ol | |
6568 | |
74.12 |
Chemical Identifiers
78-92-2 | |
74.12 | |
BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
6568 | |
butan-2-ol |
C4H10O | |
MFCD00004569 | |
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
CHEBI:35687 | |
CCC(C)O |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H226-H319-H335-H336
DOTInformation : DOT Class: 3; Packaging Group: III
EINECSNumber : 201-158-5
RTECSNumber : EO1750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only