missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butanol, anhydrous, 99%, Alfa Aesar™

Hygroscopic; H2O <0.01%

Manufacturer:  Alfa Aesar 43315K2

 View more versions of this product

Catalog No. AA43315K2



Specifications

Specifications

2-Butanol
78-92-2
99%
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYSA-N
butan-2-ol
6568
74.12
0.808g/cm3 at 20°C
Alcohol-like
Colorless
H2O <0.01%
Colorless liquid
C4H10O
1-methyl-1-propanol
CCC(C)O
74.123
CHEBI:35687
Liquid
390°C (734°F)
17.3 hPa (13mm Hg) at 20°C
1L

Chemical Identifiers

78-92-2
74.123
BTANRVKWQNVYAZ-UHFFFAOYSA-N
6568
butan-2-ol
C4H10O
MFCD00004569
1-methyl-1-propanol
CHEBI:35687
CCC(C)O
Safety and Handling

Safety and Handling

H226-H319-H335-H336: Flammable liquid and vapour. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 201-158-5

Documents