missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromovaleric Acid, TCI America™

$48.89 - $420.14

Chemical Identifiers

CAS 584-93-0
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.029
MDL Number MFCD00004217
InChI Key WMFATTFQNRPXBQ-UHFFFAOYSA-N
Synonym 2-bromovaleric acid, pentanoic acid, 2-bromo, .alpha.-bromovaleric acid, valeric acid, 2-bromo, alpha-bromo-n-valeric acid, .alpha.-bromopentanoic acid, valeric acid, .alpha.-bromo, alpha-bromovaleric acid, alpha-bromopentanoic acid, valeric acid, alpha-bromo
PubChem CID 11446
IUPAC Name 2-bromopentanoic acid
SMILES CCCC(C(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B066725G View Documents TCI America
B066725G
25g Each for $48.89
 
B0667500G View Documents TCI America
B0667500G
500g Each for $420.14
 
Specifications

Chemical Identifiers

584-93-0
181.029
WMFATTFQNRPXBQ-UHFFFAOYSA-N
11446
CCCC(C(=O)O)Br
C5H9BrO2
MFCD00004217
2-bromovaleric acid, pentanoic acid, 2-bromo, .alpha.-bromovaleric acid, valeric acid, 2-bromo, alpha-bromo-n-valeric acid, .alpha.-bromopentanoic acid, valeric acid, .alpha.-bromo, alpha-bromovaleric acid, alpha-bromopentanoic acid, valeric acid, alpha-bromo
2-bromopentanoic acid

Specifications

136°C
25g
C5H9BrO2
3265
WMFATTFQNRPXBQ-UHFFFAOYSA-N
2-bromopentanoic acid
11446
Clear Liquid at 20°C
2-Bromovaleric Acid
Yellow
584-93-0
MFCD00004217
2-bromovaleric acid, pentanoic acid, 2-bromo, .alpha.-bromovaleric acid, valeric acid, 2-bromo, alpha-bromo-n-valeric acid, .alpha.-bromopentanoic acid, valeric acid, .alpha.-bromo, alpha-bromovaleric acid, alpha-bromopentanoic acid, valeric acid, alpha-bromo
CCCC(C(=O)O)Br
181.029
181.03
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : Yes