Please login to your online account to display your discounted pricing

2-Bromovaleric acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromovaleric acid
Name Note 98%
CAS 584-93-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H9BrO2
Linear Formula CH3CH2CH2CH(Br)CO2H
Formula Weight 181.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305230250

  SDS
thermo fisher scientific
AC305230250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 181.03
Quantity 25g
Color Clear pale yellow
EINECS Number 209-546-6
TSCA TSCA
Density 1.3810g/mL
CAS Max % 100.0
Chemical Name or Material 2-Bromovaleric acid
Assay Percent Range 98%
Molecular Formula C5H9BrO2
Linear Formula CH3CH2CH2CH(Br)CO2H
CAS 584-93-0
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4700 to 1.4720 (20°C, 589nm)
Name Note 98%
MDL Number MFCD00004217
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.