Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromovaleric acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromovaleric acid
CAS 584-93-0
Molecular Formula C5H9BrO2
Formula Weight 181.03
MDL Number MFCD00004217
UN Number UN3265
Beilstein 1721172
Synonym 2-Bromopentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0704714

  SDS
alfa aesar™
L0704714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.03
Health Hazard 1 Danger
Chemical Name or Material 2-Bromovaleric acid
CAS 584-93-0
Beilstein 1721172
Synonym 2-Bromopentanoic acid
MDL Number MFCD00004217
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C5H9BrO2
UN Number UN3265
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
TSCA TSCA
Boiling Point 132° to 136°C (25mmHg)
Quantity 25g
Assay 98%
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.