Please login to your online account to display your discounted pricing

2-Bromotoluene, 99%, Alfa Aesar™

$57.73 - $57.73

Chemical Identifiers

Chemical Name or Material 2-Bromotoluene
CAS 95-46-5
Molecular Formula C7H7Br
Formula Weight 171.04
MDL Number MFCD00000068
Beilstein 1904176
Merck Index 14,1439
Synonym 2-Methylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1231514

 
alfa aesar
A1231514
Each for $57.73
Description & Specifications

Specifications

Formula Weight 171.04
Quantity 25g
TSCA TSCA
Assay 99%
Molecular Formula C7H7Br
CAS 95-46-5
Beilstein 1904176
Melting Point −27°C
Boiling Point 179° to 181°C
Flash Point 78°C (172°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335-H227;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XS7965500
Chemical Name or Material 2-Bromotoluene
MDL Number MFCD00000068
Merck Index 14,1439
Synonym 2-Methylbromobenzene

Applications
2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

Solubility
Soluble in alcohol, ether, acetone, and ethanol. Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335-H227;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.