missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromotoluene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107470250

 View more versions of this product

Catalog No. AC107470250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromotoluene
95-46-5
100.0
Colorless to Yellow
181.5°C
Authentic
Glass bottle
1.5550 to 1.557
MFCD00000068
05, 304
14, 1439
Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and benzene
CC1=CC=CC=C1Br
171.04
171.04
Liquid
99%
98.5
-27°C
1.4200g/mL
78°C
99%
C7H7Br
CH3C6H4Br
25 mL
1.42
2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
QSSXJPIWXQTSIX-UHFFFAOYSA-N
1-bromo-2-methylbenzene
7236
99%

Chemical Identifiers

95-46-5
171.04
QSSXJPIWXQTSIX-UHFFFAOYSA-N
7236
CC1=CC=CC=C1Br
C7H7Br
MFCD00000068
2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
1-bromo-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-421-7

RTECSNumber : XS7965500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only