Please login to your online account to display your discounted pricing

2-Bromothiophenol, 97%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC221980500

 View more versions of this product

Catalog No. AC221980500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Liquid
Formula Weight 189.07
Quantity 50g
Color Clear colorless to slightly yellow
Density 1.604g/mL
Beilstein 06, II, 300
CAS 1614404
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 2-Bromothiophenol
Assay Percent Range 97%
Molecular Formula C6H5BrS
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Fatal if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004827
Refractive Index 1.6340 to 1.6370 (20°C, 589nm)
Synonym 2-Bromobenzenethiol
Linear Formula BrC6H4SH

Specifications

Chemical Name or Material 2-Bromothiophenol
CAS 1614404
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H5BrS
Linear Formula BrC6H4SH
Formula Weight 189.07
MDL Number MFCD00004827
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Fatal if inhaled.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.

SDS & Safety Documents