missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H5523803

 View more versions of this product

Catalog No. AAH5523803



Specifications

Specifications

2-Bromothiobenzamide
82°C to 86°C
30216-44-5
MFCD04973314
GHCQWSAFBXLYSO-UHFFFAOYSA-N
2-bromobenzenecarbothioamide
2734818
Solid
Brown to White
1g
C7H6BrNS
2-bromo-thiobenzamide
C1=CC=C(C(=C1)C(=S)N)Br
216.096
216.1
97%

Chemical Identifiers

30216-44-5
216.096
GHCQWSAFBXLYSO-UHFFFAOYSA-N
2734818
C1=CC=C(C(=C1)C(=S)N)Br
C7H6BrNS
MFCD04973314
2-bromo-thiobenzamide
2-bromobenzenecarbothioamide
Documents