Please login to your online account to display your discounted pricing

2-Bromothioanisole, 98%, ACROS Organics™

$42.64 - $42.64

Chemical Identifiers

Chemical Name or Material 2-Bromothioanisole
CAS 19614-16-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7BrS
Linear Formula BrC6H4SCH3
Formula Weight 203.11
MDL Number MFCD00000066
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303610010

  SDS
thermo fisher scientific
303610010
Glass bottle 1g Each for $42.64
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 203.11
Quantity 1g
Color Clear colorless to yellow
CAS Max % 100.0
Chemical Name or Material 2-Bromothioanisole
Assay Percent Range 98%
Synonym 2-Bromophenyl methyl sulfide
Molecular Formula C7H7BrS
CAS 19614-16-5
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00000066
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula BrC6H4SCH3
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.6320 to 1.6340 (20°C, 589nm)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.