missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromothioanisole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 303610010

 View more versions of this product

Catalog No. AC303610010


 

Specifications Description & Specifications

Specifications

≥97.5%
203.11
Clear colorless to yellow
2-Bromophenyl methyl sulfide
98%
19614-16-5
MFCD00000066
Glass bottle
BrC6H4SCH3
Liquid
1g
100.0
2-Bromothioanisole
C7H7BrS
97.5
1.6320 to 1.6340 (20°C, 589nm)
Authentic

Chemical Identifiers

19614-16-5
100.0
C7H7BrS
203.11
2-Bromophenyl methyl sulfide
≥97.5%
97.5
98%
BrC6H4SCH3
MFCD00000066
2-Bromothioanisole

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning