Learn More
2-Bromothiazole-4-carboxaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 429540050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Bromothiazole-4-carboxaldehyde | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD04115726 | |
2-bromothiazole-4-carbaldehyde, 2-bromo-4-formylthiazole, 2-bromo-4-fomylthiazole, 2-bromothiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-bromo, 2-bromo-1,3-thiazole-4-carboxaldehyde, 2-bromo-4-formyl-1,3-thiazole, pubchem8915, acmc-209kx4, 2-bromo-4-thiazole carboxaldehyde | |
BrC1=CSC(C=O)=N1 | |
192.03 | |
192.03 |
5198-80-1 | |
100.0 | |
96% min. (HPLC) | |
C4H2BrNOS | |
5g | |
JDUXMFGFGCJNGO-UHFFFAOYSA-N | |
2-bromo-1,3-thiazole-4-carbaldehyde | |
17750433 | |
97% |
Chemical Identifiers
5198-80-1 | |
192.03 | |
JDUXMFGFGCJNGO-UHFFFAOYSA-N | |
17750433 | |
BrC1=CSC(C=O)=N1 |
C4H2BrNOS | |
MFCD04115726 | |
2-bromothiazole-4-carbaldehyde, 2-bromo-4-formylthiazole, 2-bromo-4-fomylthiazole, 2-bromothiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-bromo, 2-bromo-1,3-thiazole-4-carboxaldehyde, 2-bromo-4-formyl-1,3-thiazole, pubchem8915, acmc-209kx4, 2-bromo-4-thiazole carboxaldehyde | |
2-bromo-1,3-thiazole-4-carbaldehyde |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO â Research Use Only