Please login to your online account to display your discounted pricing

2-Bromoterephthalic acid, 95%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC369811000

 View more versions of this product

Catalog No. AC369811000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 245.03
Quantity 100g
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C8H5BrO4
Linear Formula BrC6H3(COH)
CAS 586-35-6
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Bromoterephthalic acid
Name Note 95%
Assay Percent Range 95%
MDL Number MFCD00002403

Specifications

Chemical Name or Material 2-Bromoterephthalic acid
Name Note 95%
CAS 586-35-6
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H5BrO4
Linear Formula BrC6H3(COH)
Formula Weight 245.03
MDL Number MFCD00002403
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents