Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoquinoline, 96%

$118.14 - $1,056.24

Chemical Identifiers

Chemical Name or Material 2-Bromoquinoline
CAS 2005-43-8
Molecular Formula C9H6BrN
Formula Weight 208.06
MDL Number MFCD00234480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2730203

  SDS
alfa aesar™
H2730203
1g Each for $118.14

AAH2730206

  SDS
alfa aesar™
H2730206
5g Each for $293.82

AAH2730214

  SDS
alfa aesar™
H2730214
25g Each for $1,056.24
Description & Specifications

Specifications

Formula Weight 208.06
Chemical Name or Material 2-Bromoquinoline
Health Hazard 1 Warning
Assay 96%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00234480
Quantity 1g
Molecular Formula C9H6BrN
CAS 2005-43-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.