missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoquinoline-3-boronic acid, 97%

$121.15 - $1,351.49

Chemical Identifiers

CAS 745784-05-8
Molecular Formula C9H7BBrNO2
Formula Weight 251.88
MDL Number MFCD05664125
Chemical Name or Material 2-Bromoquinoline-3-boronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2032703 SDS Alfa Aesar™
L2032703
1g Each for $349.42
AAL2032706 SDS Alfa Aesar™
L2032706
5g Each for $1,351.49
AAL20327MD SDS Alfa Aesar™
L20327MD
250mg Each for $121.15
Specifications Description & Specifications

Chemical Identifiers

745784-05-8
251.88
2-Bromoquinoline-3-boronic acid
C9H7BBrNO2
MFCD05664125

Specifications

745784-05-8
251.88
144° to 147°C
2-Bromoquinoline-3-boronic acid
C9H7BBrNO2
MFCD05664125
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning