Learn More
2-Bromopyridine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 107401000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromopyridine | |
98.5 | |
Colorless to Brown | |
192°C to 194°C (761.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5705 to 1.5725 | |
MFCD00006219 | |
20, 233 | |
o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8 | |
IMRWILPUOVGIMU-UHFFFAOYSA-N | |
2-bromopyridine | |
7973 | |
158 | |
Liquid |
109-04-6 | |
100.0 | |
1.6500g/mL | |
54°C | |
98.5% min. (GC) | |
C5H4BrN | |
(C5H4N)Br | |
100 mL | |
1.65 | |
Solubility in water: 2.5g/L water (20°C) | |
C1=CC=NC(=C1)Br | |
158 | |
CHEBI:51574 | |
99% |
Chemical Identifiers
109-04-6 | |
158 | |
IMRWILPUOVGIMU-UHFFFAOYSA-N | |
7973 | |
2-bromopyridine |
C5H4BrN | |
MFCD00006219 | |
o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8 | |
CHEBI:51574 | |
C1=CC=NC(=C1)Br |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date
GHS Signal Word: Danger
EINECSNumber : 203-641-6
RUO – Research Use Only