Learn More
2-Bromopyridine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1324114
Description
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.
Solubility
Slightly miscible with water.
Notes
Keep away from heat, sparks, and flame. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and acid chlorides.
Specifications
2-Bromopyridine | |
1.625 | |
97°C (207°F) | |
C5H4BrN | |
MFCD00006219 | |
UN2810 | |
o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8 | |
IMRWILPUOVGIMU-UHFFFAOYSA-N | |
2-bromopyridine | |
7973 | |
158 |
109-04-6 | |
192°C to 194°C | |
Characteristic | |
1.572 | |
25 g | |
105789 | |
Slightly miscible with water. | |
C1=CC=NC(=C1)Br | |
157.998 | |
CHEBI:51574 | |
99% |
Chemical Identifiers
109-04-6 | |
157.998 | |
IMRWILPUOVGIMU-UHFFFAOYSA-N | |
7973 | |
2-bromopyridine |
C5H4BrN | |
MFCD00006219 | |
o-bromopyridine, pyridine, 2-bromo, 2-bromo-pyridine, 2-pyridyl bromide, alpha-bromopyridine, beta-bromopyridine, bromopyridine, 2-bromopyridin, 2-bromo pyridine, unii-7z7mlc4vd8 | |
CHEBI:51574 | |
C1=CC=NC(=C1)Br |
Safety and Handling
GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P362-P374-P380-P501c
H226-H301+H331-H310-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 203-641-6
RTECSNumber : US3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only