Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromopyridine-5-boronic acid pinacol ester, 98%

$281.77 - $1,240.07

Chemical Identifiers

Chemical Name or Material 2-Bromopyridine-5-boronic acid pinacol ester
CAS 214360-62-0
Molecular Formula C11H15BBrNO2
Formula Weight 283.96
MDL Number MFCD03412794
Synonym 2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5007203

  SDS
alfa aesar™
H5007203
1g Each for $281.77

AAH5007206

  SDS
alfa aesar™
H5007206
5g Each for $1,240.07
Description & Specifications

Specifications

Formula Weight 283.96
Assay 98%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Chemical Name or Material 2-Bromopyridine-5-boronic acid pinacol ester
MDL Number MFCD03412794
CAS 214360-62-0
Molecular Formula C11H15BBrNO2
Synonym 2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.