Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromopyridine-4-carboxaldehyde, 97%

$148.72 - $604.22

Chemical Identifiers

Chemical Name or Material 2-Bromopyridine-4-carboxaldehyde
CAS 118289-17-1
Molecular Formula C6H4BrNO
Formula Weight 186.01
MDL Number MFCD04039311
Beilstein 4243166
Synonym 2-Bromoisonicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1985206

  SDS
alfa aesar™
L1985206
5g Each for $604.22

AAL1985203

  SDS
alfa aesar™
L1985203
1g Each for $148.72
Description & Specifications

Specifications

Chemical Name or Material 2-Bromopyridine-4-carboxaldehyde
CAS 118289-17-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage 2° to 8°C
Assay 97%
Molecular Formula C6H4BrNO
Formula Weight 186.01
MDL Number MFCD04039311
Melting Point 52° to 56°C
Beilstein 4243166
Synonym 2-Bromoisonicotinaldehyde
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C