missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromopyridine-4-boronic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435920050

 View more versions of this product

Catalog No. AC435920050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromopyridine-4-boronic acid
White
458532-94-0
100.0
C5H5BBrNO2
DFIKFQMGZNTFLI-UHFFFAOYSA-N
(2-bromopyridin-4-yl)boronic acid
12060162
Crystalline Powder
Authentic
5g
94.0
94% min. (HPLC)
2-bromanylpyridin-4-yl boronic acid, 2-bromo-4-pyridineboronic acid, 2-bromo-4-pyridinyl boronic acid, 2-bromo-4-pyridyl boronic acid, 2-bromopyridin-4-yl boronic acid, 2-bromopyridine-4-boronic acid, acmc-209k4o, boronic acid, 2-bromo-4-pyridinyl-9ci, boronicacid, 2-bromo-4-pyridinyl-9ci, ksc234s1f
B(C1=CC(=NC=C1)Br)(O)O
201.82
201.82
95%

Chemical Identifiers

458532-94-0
201.82
2-bromanylpyridin-4-yl boronic acid, 2-bromo-4-pyridineboronic acid, 2-bromo-4-pyridinyl boronic acid, 2-bromo-4-pyridyl boronic acid, 2-bromopyridin-4-yl boronic acid, 2-bromopyridine-4-boronic acid, acmc-209k4o, boronic acid, 2-bromo-4-pyridinyl-9ci, boronicacid, 2-bromo-4-pyridinyl-9ci, ksc234s1f
(2-bromopyridin-4-yl)boronic acid
C5H5BBrNO2
DFIKFQMGZNTFLI-UHFFFAOYSA-N
12060162
B(C1=CC(=NC=C1)Br)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents