Please login to your online account to display your discounted pricing

2-Bromopyridine-3-carboxylic acid, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC429770010

 View more versions of this product

Catalog No. AC429770010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 202.01
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
MDL Number MFCD01646068
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 2-Bromopyridine-3-carboxylic acid
CAS 35905-85-2
Molecular Formula C6H4BrNO2
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Bromopyridine-3-carboxylic acid
CAS 35905-85-2
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H4BrNO2
Formula Weight 202.01
MDL Number MFCD01646068
Accessories

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

SDS & Safety Documents