missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromopropiophenone, TCI America™

$119.17 - $695.16

Chemical Identifiers

CAS 2114-00-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000145
InChI Key WPDWOCRJBPXJFM-UHFFFAOYSA-N
Synonym 2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
PubChem CID 16452
IUPAC Name 2-bromo-1-phenylpropan-1-one
SMILES CC(C(=O)C1=CC=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B293325G View Documents TCI America
B293325G
25g Each for $119.17
 
B2933250G View Documents TCI America
B2933250G
250g Each for $695.16
 
Specifications

Chemical Identifiers

2114-00-3
213.074
WPDWOCRJBPXJFM-UHFFFAOYSA-N
16452
CC(C(=O)C1=CC=CC=C1)Br
C9H9BrO
MFCD00000145
2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
2-bromo-1-phenylpropan-1-one

Specifications

137°C
25g
C9H9BrO
2810
WPDWOCRJBPXJFM-UHFFFAOYSA-N
2-bromo-1-phenylpropan-1-one
16452
Clear Liquid at 20°C
2-Bromopropiophenone
Undesignated
2114-00-3
MFCD00000145
2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
CC(C(=O)C1=CC=CC=C1)Br
213.074
213.07
≥96.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : UG8400000

TSCA : Yes

Recommended Storage : Refrigerator