missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromopropiophenone 96.0+%, TCI America™

Manufacturer:  TCI America B2933250G

 View more versions of this product

Catalog No. B2933250G



Specifications

Specifications

2-Bromopropiophenone
Undesignated
2114-00-3
MFCD00000145
2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
CC(C(=O)C1=CC=CC=C1)Br
213.074
213.07
≥96.0% (GC)
137°C
250g
C9H9BrO
2810
WPDWOCRJBPXJFM-UHFFFAOYSA-N
2-bromo-1-phenylpropan-1-one
16452
Clear Liquid at 20°C

Chemical Identifiers

2114-00-3
213.074
WPDWOCRJBPXJFM-UHFFFAOYSA-N
16452
CC(C(=O)C1=CC=CC=C1)Br
C9H9BrO
MFCD00000145
2-bromopropiophenone, 1-propanone, 2-bromo-1-phenyl, alpha-bromopropiophenone, 1-benzoyl-1-bromoethane, 1-bromoethyl phenyl ketone, 2-bromo-1-phenyl-1-propanone, propiophenone, 2-bromo, .alpha.-bromopropiophenone, 2-bromo-1-phenyl-propan-1-one, 2-bromo-propiophenone
2-bromo-1-phenylpropan-1-one
Safety and Handling

Safety and Handling

RTECSNumber : UG8400000

TSCA : Yes

Recommended Storage : Refrigerator

Documents