Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromopropionic acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromopropionic acid
CAS 598-72-1
Molecular Formula CH3CHBrCO2H
Formula Weight 152.98
MDL Number MFCD00004211
UN Number UN3261
Beilstein 605451
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1756930

  SDS
alfa aesar™
A1756930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.98
Chemical Name or Material 2-Bromopropionic acid
TSCA TSCA
Melting Point 25°C
Boiling Point 202° to 203°C
Quantity 250g
Flash Point 100°C (212°F)
Assay 98%
Molecular Formula CH3CHBrCO2H
MDL Number MFCD00004211
CAS 598-72-1
UN Number UN3261
Beilstein 605451
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number UA2451715

Applications
It is employed as pharmaceutical and agrochemical intermediate. It is used as an alkylation agent for mercaptans and other sulfur containing compounds.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.