missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Bromophenylethynyl)trimethylsilane, 98%

Manufacturer:  Alfa Aesar™ H5340203

 View more versions of this product

Catalog No. AAH5340203



Description

Description

Applications
(2-Bromophenylethynyl)trimethylsilane finds wide range application in the medicine and electronic industry. Reaction of (2-bromophenylethynyl)trimethylsilane 1 with trimethyl borate yielded the alkynearylboronic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

(2-Bromophenylethynyl)trimethylsilane
C11H13BrSi
2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, 2-bromophenylethynyl trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl
C[Si](C)(C)C#CC1=CC=CC=C1Br
253.214
253.21
60°C to 80°C (2mmHg)
>113°C (235°F)
38274-16-7
MFCD01321391
FABNGXSCLHXUOH-UHFFFAOYSA-N
2-(2-bromophenyl)ethynyl-trimethylsilane
4547398
98%
1g

Chemical Identifiers

38274-16-7
253.214
FABNGXSCLHXUOH-UHFFFAOYSA-N
4547398
C[Si](C)(C)C#CC1=CC=CC=C1Br
C11H13BrSi
MFCD01321391
2-bromophenylethynyl trimethylsilane, 2-bromophenyl ethynyl trimethylsilane, 2-2-bromophenyl ethynyl trimethylsilane, acmc-20appv, 2-bromophenyl trimethylsilyl acetylene, 2-2-bromophenyl ethynyl-trimethylsilane, 2-bromophenylethynyl trimethylsilane, benzene,1-bromo-2-2-trimethylsilyl ethynyl
2-(2-bromophenyl)ethynyl-trimethylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents