missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromophenylboronic acid, 97%, Thermo Scientific Chemicals
$134.94 - $1907.60
Chemical Identifiers
CAS | 244205-40-1 |
---|---|
Molecular Formula | C6H6BBrO2 |
Molecular Weight (g/mol) | 200.83 |
MDL Number | MFCD01114672 |
InChI Key | PLVCYMZAEQRYHJ-UHFFFAOYSA-N |
Synonym | 2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid |
PubChem CID | 2773294 |
IUPAC Name | (2-bromophenyl)boronic acid |
SMILES | OB(O)C1=CC=CC=C1Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC379990050
|
Thermo Scientific Chemicals
379990050 |
5 g | Glass bottle |
Each for $134.94
|
|
||||
AC379990250
|
Thermo Scientific Chemicals
379990250 |
25 g | Glass bottle |
Each for $479.39
|
|
||||
AC379991000
|
Thermo Scientific Chemicals
379991000 |
100 g | Glass Bottle |
Each for $1,907.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
244205-40-1 | |
200.83 | |
PLVCYMZAEQRYHJ-UHFFFAOYSA-N | |
2773294 | |
OB(O)C1=CC=CC=C1Br |
C6H6BBrO2 | |
MFCD01114672 | |
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid | |
(2-bromophenyl)boronic acid |
Specifications
244205-40-1 | |
White | |
96% min. (HPLC) | |
C6H6BBrO2 | |
MFCD01114672 | |
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid | |
OB(O)C1=CC=CC=C1Br | |
200.83 | |
200.83 | |
Powder |
109.0°C to 114.0°C | |
Authentic | |
Glass bottle | |
BrC6H4B(OH)2 | |
5 g | |
PLVCYMZAEQRYHJ-UHFFFAOYSA-N | |
(2-bromophenyl)boronic acid | |
2773294 | |
97% | |
2-Bromophenylboronic acid |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only