Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromophenylacetonitrile, 97%

$16.00 - $16.00

Chemical Identifiers

Chemical Name or Material 2-Bromophenylacetonitrile
CAS 19472-74-3
Molecular Formula C8H6BrN
Formula Weight 196.05
MDL Number MFCD00001896
UN Number UN1694
Beilstein 2043684
Synonym 2-Bromobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1754806

  SDS
alfa aesar™
A1754806
5g Each for $16.00

AAA1754814

  SDS
alfa aesar™
A1754814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.05
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P361-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 2-Bromophenylacetonitrile
Assay 97%
Molecular Formula C8H6BrN
MDL Number MFCD00001896
CAS 19472-74-3
UN Number UN1694
Melting Point 19°C
Beilstein 2043684
Synonym 2-Bromobenzyl cyanide
Boiling Point 252°C
Quantity 5g
Flash Point 113°C (235°F)
Odor Pungent
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BROMOBENZYL CYANIDES, LIQUID
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P361-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.