Please login to your online account to display your discounted pricing

2-Bromophenylacetone, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  350590050

 View more versions of this product

Catalog No. AC350590050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 213.08
Quantity 5g
Color Clear colorless to yellow
TSCA TSCA
CAS Max % 100.0
Molecular Formula C9H9BrO
Infrared Spectrum Authentic
Chemical Name or Material 2-Bromophenylacetone
CAS 21906-31-0
CAS Min % 98.5
Assay Percent Range 99%
MDL Number MFCD03410431
Name Note 99%
Packaging Glass bottle
Refractive Index 1.5575 to 1.5595 (20°C, 589nm)

Specifications

Chemical Name or Material 2-Bromophenylacetone
Name Note 99%
CAS 21906-31-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H9BrO
Formula Weight 213.08
MDL Number MFCD03410431