Please login to your online account to display your discounted pricing

2-Bromophenylacetone, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350590010

 View more versions of this product

Catalog No. AC350590010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 213.08
Physical Form Liquid
Quantity 1g
Color Clear colorless to yellow
TSCA TSCA
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C9H9BrO
CAS 21906-31-0
CAS Min % 98.5
Name Note 99%
Chemical Name or Material 2-Bromophenylacetone
MDL Number MFCD03410431
Assay Percent Range 99%
Refractive Index 1.5575 to 1.5595 (20°C, 589nm)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Bromophenylacetone
Name Note 99%
CAS 21906-31-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H9BrO
Formula Weight 213.08
MDL Number MFCD03410431