missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromophenyl Trifluoromethanesulfonate, TCI America™

$87.08 - $293.34

Chemical Identifiers

CAS 129112-25-0
Molecular Formula C7H4BrF3O3S
MDL Number MFCD18379777
Synonym Trifluoromethanesulfonic Acid 2-Bromophenyl Ester, 2-Bromophenyl Triflate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B47771G View Documents TCI America
B47771G
1g Each for $87.08
 
B47775G View Documents TCI America
B47775G
5g Each for $293.34
 
Specifications

Chemical Identifiers

129112-25-0
C7H4BrF3O3S
MFCD18379777
Trifluoromethanesulfonic Acid 2-Bromophenyl Ester, 2-Bromophenyl Triflate

Specifications

113°C
1g
C7H4BrF3O3S
Trifluoromethanesulfonic Acid 2-Bromophenyl Ester, 2-Bromophenyl Triflate
Clear Liquid at 20°C
2-Bromophenyl Trifluoromethanesulfonate
Yellow
129112-25-0
MFCD18379777
305.07
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No