missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromophenyl Isocyanate 98.0+%, TCI America™
Supplier: TCI America I049825G
Specifications
2-Bromophenyl Isocyanate | |
Yellow | |
MFCD00001995 | |
2206 | |
GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
1-bromo-2-isocyanatobenzene | |
137097 | |
≥98.0% (GC) |
1592-00-3 | |
C7H4BrNO | |
25 g | |
2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
C1=CC=C(C(=C1)N=C=O)Br | |
198.019 | |
198.02 | |
Liquid |
Chemical Identifiers
1592-00-3 | |
198.019 | |
GOOVAYJIVMBWPP-UHFFFAOYSA-N | |
137097 | |
C1=CC=C(C(=C1)N=C=O)Br |
C7H4BrNO | |
MFCD00001995 | |
2-bromophenyl isocyanate, 2-bromophenylisocyanate, benzene, 1-bromo-2-isocyanato, isocyanic acid 2-bromophenyl ester, 2-bromobenzenisocyanate, 2bromophenylisocyanate, 2-bromophenylisocynate, o-bromophenyl isocyanate, acmc-209djl, 2-bromo phenylisocyanate | |
1-bromo-2-isocyanatobenzene |
Safety and Handling
TSCA : No